Assay Details
| Gene Symbol: G6pc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008061.3 | NP_032087.2 | 4 - 5 | 623 | 112 | |
| GenBank mRNA | AK050279.1 | - | 4 - 5 | 638 | 112 | |
| AK052656.1 | - | 4 - 5 | 638 | 112 | ||
| BC013448.1 | - | 4 - 5 | 623 | 4237115 | 112 | |
| U00445.1 | - | 4 - 5 | 638 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycogen metabolic process
glycogen catabolic process
gluconeogenesis
triglyceride metabolic process
phosphate-containing compound metabolic process
steroid metabolic process
regulation of gene expression
glucose-6-phosphate transport
dephosphorylation
multicellular organism growth
glucose homeostasis
cholesterol homeostasis
urate metabolic process
phosphorylated carbohydrate dephosphorylation
glucose 6-phosphate metabolic process
lipid homeostasis
catalytic activity
glucose-6-phosphatase activity
phosphotransferase activity, alcohol group as acceptor
hydrolase activity
phosphate ion binding
glycogen metabolic process
glycogen catabolic process
gluconeogenesis
triglyceride metabolic process
phosphate-containing compound metabolic process
steroid metabolic process
regulation of gene expression
glucose-6-phosphate transport
dephosphorylation
multicellular organism growth
glucose homeostasis
cholesterol homeostasis
urate metabolic process
phosphorylated carbohydrate dephosphorylation
glucose 6-phosphate metabolic process
lipid homeostasis
catalytic activity
glucose-6-phosphatase activity
phosphotransferase activity, alcohol group as acceptor
hydrolase activity
phosphate ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462385 | U00445.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P462385 | U00445.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673649 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334074 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22117 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90938 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 171235 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 941356 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157154 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145156 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462385 | U00445.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P462385 | U00445.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P462385 | U00445.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P462385 | U00445.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073554 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28073544 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145156 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07466499 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157154 | - | - |
| Whole Mouse Genome G4122A | A_51_P462385 | U00445.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P462385 | U00445.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101020 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139061 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145156 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157154 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09313194 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09212220 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09212218 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09212221 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09212216 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09212217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28073548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09479201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09264824 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09701104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09172206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09191879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09479196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09212220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073572 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09212221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09338548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09723765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09172215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09619686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09268291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09212216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09052323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28073513 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10381393 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381394 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417880_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10381387 | - | - |
| Mouse Genome 430 2.0 Array | 1417880_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417880_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14377_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14377_PM_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa1190_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100015498.mm.1 | NM_008061.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015498.mm.1 | TC1100001654.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417880_PM_at | - | - |
| Mouse Expression Set 430 | 1417880_at | - | - |
| Murine Genome U74v2 Set | 103333_PM_at | - | - |
| Murine Genome U74v2 Set | 103333_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417880_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417880_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417880_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417880_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 333144 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 430093 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 430093 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000430093 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245103 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1245103 | - | - |
| mouse-6 v1.1 (Illumina) | scl40883.5_194-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 430093 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40883.5_194-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430093 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1245103 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245103 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40883.5_194-S | - | - |

G6pc
Genomic Map