Assay Details
| Gene Symbol: Gad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008077.4 | NP_032103.2 | 11 - 12 | 1352 | 101 | |
| GenBank mRNA | AF326547.1 | - | 11 - 12 | 1302 | 101 | |
| AF326548.1 | - | 11 - 12 | 1302 | 101 | ||
| AF483492.1 | - | 10 - 11 | 1119 | 101 | ||
| AF483493.1 | - | 10 - 11 | 1119 | 101 | ||
| AK136801.1 | - | 11 - 12 | 1337 | 101 | ||
| AK148497.1 | - | 11 - 12 | 1352 | 101 | ||
| AK158827.1 | - | 10 - 11 | 1118 | 101 | ||
| AK160410.1 | - | 10 - 11 | 1261 | 101 | ||
| AK160868.1 | - | 11 - 12 | 1350 | 101 | ||
| BC027059.1 | - | 11 - 12 | 1303 | 5358787 | 101 | |
| KC134210.1 | - | 10 - 11 | 1119 | 101 | ||
| S67453.1 | - | 6 - 7 | 528 | 101 | ||
| Y12257.1 | - | 10 - 11 | 1119 | 101 | ||
| Z49976.1 | - | 11 - 12 | 1239 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gad1
Genomic Map