Assay Details
| Gene Symbol: Gad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008077.4 | NP_032103.2 | 15 - 16 | 1745 | 113 | |
| GenBank mRNA | AF326547.1 | - | 15 - 16 | 1695 | 113 | |
| AF326548.1 | - | 15 - 16 | 1695 | 113 | ||
| AF483492.1 | - | 14 - 15 | 1512 | 113 | ||
| AF483493.1 | - | 14 - 15 | 1512 | 113 | ||
| AK136801.1 | - | 15 - 16 | 1730 | 113 | ||
| AK148497.1 | - | 15 - 16 | 1745 | 113 | ||
| AK158827.1 | - | 14 - 15 | 1511 | 113 | ||
| AK160410.1 | - | 14 - 15 | 1654 | 113 | ||
| AK160868.1 | - | 15 - 16 | 1743 | 113 | ||
| AK163893.1 | - | 1 - 2 | 90 | 113 | ||
| BC027059.1 | - | 15 - 16 | 1696 | 5358787 | 113 | |
| KC134210.1 | - | 14 - 15 | 1512 | 113 | ||
| Y12257.1 | - | 14 - 15 | 1512 | 113 | ||
| Z49976.1 | - | 15 - 16 | 1632 | 113 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gad1
Genomic Map