Assay Details
| Gene Symbol: Gad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008077.4 | NP_032103.2 | 1 - 2 | 171 | 107 | |
| GenBank mRNA | AF326547.1 | - | 1 - 2 | 121 | 107 | |
| AF326548.1 | - | 1 - 2 | 121 | 107 | ||
| AK054554.1 | - | 1 - 2 | 173 | 107 | ||
| AK136801.1 | - | 1 - 2 | 156 | 107 | ||
| AK148497.1 | - | 1 - 2 | 171 | 107 | ||
| AK160868.1 | - | 1 - 2 | 169 | 107 | ||
| BC027059.1 | - | 1 - 2 | 122 | 5358787 | 107 | |
| KM102984.1 | - | 1 - 2 | 171 | 107 | ||
| Z49976.1 | - | 1 - 2 | 58 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF326547.1, AF326548.1, AK054554.1, AK136801.1, AK148497.1, AK160868.1, BC027059.1, KM102984.1, NM_008077.4, Z49976.1 transcripts.
Set Membership: |
|

Gad1
Genomic Map