Assay Details
| Gene Symbol: Mcm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008568.2 | NP_032594.1 | 3 - 4 | 381 | 114 | |
| GenBank mRNA | AK132748.1 | - | 942 | 114 | ||
| AK133091.1 | - | 3 - 4 | 377 | 114 | ||
| AK144980.1 | - | 3 - 4 | 393 | 114 | ||
| AK154052.1 | - | 3 - 4 | 378 | 114 | ||
| BC023232.1 | - | 3 - 4 | 357 | 114 | ||
| BC065164.1 | - | 3 - 4 | 365 | 6829737 | 114 | |
| BC066024.1 | - | 3 - 4 | 320 | 6829337 | 114 | |
| D26091.1 | - | 3 - 4 | 381 | 114 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mcm7
Genomic Map