Assay Details
| Gene Symbol: Mcm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008568.2 | NP_032594.1 | 4 - 5 | 503 | 90 | |
| GenBank mRNA | AK077766.1 | - | 1 - 2 | 66 | 90 | |
| AK132748.1 | - | 1064 | 90 | |||
| AK133091.1 | - | 4 - 5 | 499 | 90 | ||
| AK144980.1 | - | 4 - 5 | 515 | 90 | ||
| AK150059.1 | - | 4 - 5 | 500 | 90 | ||
| AK154052.1 | - | 4 - 5 | 500 | 90 | ||
| BC023232.1 | - | 4 - 5 | 479 | 90 | ||
| BC065164.1 | - | 4 - 5 | 487 | 6829737 | 90 | |
| BC066024.1 | - | 4 - 5 | 442 | 6829337 | 90 | |
| D26091.1 | - | 4 - 5 | 503 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mcm7
Genomic Map