Assay Details
| Gene Symbol: Mcm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008568.2 | NP_032594.1 | 9 - 10 | 1219 | 100 | |
| GenBank mRNA | AK076059.1 | - | 5 - 6 | 669 | 100 | |
| AK133091.1 | - | 9 - 10 | 1215 | 100 | ||
| AK144980.1 | - | 9 - 10 | 1231 | 100 | ||
| AK150059.1 | - | 9 - 10 | 1216 | 100 | ||
| AK154052.1 | - | 9 - 10 | 1216 | 100 | ||
| BC023232.1 | - | 9 - 10 | 1195 | 100 | ||
| BC065164.1 | - | 9 - 10 | 1203 | 6829737 | 100 | |
| BC066024.1 | - | 9 - 10 | 1158 | 6829337 | 100 | |
| D26091.1 | - | 9 - 10 | 1219 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mcm7
Genomic Map