Assay Details
| Gene Symbol: Mt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008631.2 | NP_032657.1 | 2 - 3 | 134 | 89 | |
| GenBank mRNA | AK131894.1 | - | 2 - 3 | 134 | 89 | |
| BC099850.1 | - | 2 - 3 | 93 | 89 | ||
| BC099851.1 | - | 2 - 3 | 94 | 89 | ||
| BC099852.1 | - | 2 - 3 | 94 | 89 | ||
| BC099853.1 | - | 2 - 3 | 93 | 89 | ||
| BC111805.1 | - | 2 - 3 | 87 | 89 | ||
| BC141870.1 | - | 2 - 3 | 95 | 40038882 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mt4
Genomic Map