Assay Details
| Gene Symbol: Pip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008843.3 | NP_032869.2 | 1 - 2 | 119 | 117 | |
| GenBank mRNA | AB017907.1 | - | 1 - 2 | 97 | 117 | |
| AK138795.1 | - | 1 - 2 | 120 | 117 | ||
| BC018264.1 | - | 1 - 2 | 83 | 117 | ||
| BC028889.1 | - | 1 - 2 | 117 | 4165968 | 117 | |
| S73282.1 | - | 1 - 2 | 115 | 117 | ||
| X02510.1 | - | 1 - 2 | 114 | 116 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the X02510.1 transcripts.
Set Membership: |
|

Pip
Genomic Map