Assay Details
| Gene Symbol: Npy4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008919.4 | NP_032945.3 | 1 - 2 | 158 | 79 | |
| GenBank mRNA | AK035505.1 | - | 229 | 79 | ||
| AK144461.1 | - | 209 | 79 | |||
| BC132395.1 | - | 1 - 2 | 158 | 40130672 | 79 | |
| BC145981.1 | - | 1 - 2 | 158 | 40131284 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK035505.1, AK144461.1, BC132395.1, BC145981.1, NM_008919.4 transcripts.
Set Membership: |
|

Npy4r
Genomic Map