Assay Details
| Gene Symbol: Prm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008933.1 | NP_032959.1 | 1 - 2 | 360 | 98 | |
| GenBank mRNA | AK005729.1 | - | 1 - 2 | 369 | 98 | |
| BC049612.1 | - | 1 - 2 | 365 | 6774072 | 98 | |
| M16456.1 | - | 1 - 2 | 263 | 98 | ||
| M27501.1 | - | 1 - 2 | 263 | 98 | ||
| X14004.1 | - | 1 - 2 | 360 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prm2
Genomic Map