Assay Details
| Gene Symbol: Tcp10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011553.4 | NP_035683.2 | 9 - 10 | 1189 | 95 | |
| NR_046291.1 | 9 - 10 | 1341 | 95 | |||
| GenBank mRNA | AK161455.1 | - | 9 - 10 | 1160 | 95 | |
| BC050771.1 | - | 9 - 10 | 6773681 | 95 | ||
| BC052353.1 | - | 9 - 10 | 1304 | 6773668 | 95 | |
| M22600.1 | - | 8 - 9 | 95 | |||
| X58169.1 | - | 9 - 10 | 95 | |||
| X58171.1 | - | 9 - 10 | 95 | |||
| Gene Symbol: Tcp10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009341.2 | NP_033367.2 | 9 - 10 | 95 | ||
| GenBank mRNA | AK077165.1 | - | 9 - 10 | 95 | ||
| BC141264.1 | - | 8 - 9 | 9055876 | 95 | ||
| M22599.1 | - | 8 - 9 | 95 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Tcp10a
Genomic Map