Assay Details
| Gene Symbol: Ccr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009916.2 | NP_034046.2 | 1 - 2 | 53 | 78 | |
| GenBank mRNA | AK042013.1 | - | 1 - 2 | 53 | 78 | |
| BC117041.1 | - | 1 - 2 | 59 | 40126360 | 78 | |
| BC119171.1 | - | 1 - 2 | 58 | 8733920 | 78 | |
| X90862.1 | - | 1 - 2 | 81 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC117041.1, BC119171.1, NM_009916.2, X90862.1 transcripts.
Set Membership: |
|

Ccr4
Genomic Map