Assay Details
| Gene Symbol: Ccr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009917.5 | NP_034047.2 | 1 - 2 | 93 | 67 | |
| GenBank mRNA | AK141906.1 | - | 1 - 2 | 64 | 67 | |
| AK154595.1 | - | 1 - 2 | 94 | 67 | ||
| AK155628.1 | - | 1 - 2 | 67 | 67 | ||
| AK155632.1 | - | 1 - 2 | 64 | 67 | ||
| AK155867.1 | - | 1 - 2 | 58 | 67 | ||
| AK170303.1 | - | 1 - 2 | 64 | 67 | ||
| BC103574.1 | - | 1 - 2 | 32 | 40045779 | 67 | |
| BC103586.1 | - | 1 - 2 | 32 | 40045781 | 67 | |
| BC103587.1 | - | 1 - 2 | 32 | 40045780 | 67 | |
| BC105643.1 | - | 1 - 2 | 33 | 40045777 | 67 | |
| BY198155.1 | - | 1 - 2 | 94 | 67 | ||
| D83648.1 | - | 1 - 2 | 63 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK141906.1, AK154595.1, AK155628.1, AK155867.1, AK170303.1, BC103574.1, BC103586.1, BC103587.1, BC105643.1, D83648.1, NM_009917.5 transcripts.
Set Membership: |
|

Ccr5
Genomic Map