Assay Details
| Gene Symbol: Hoxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010449.4 | NP_034579.3 | 1 - 2 | 755 | 103 | |
| GenBank mRNA | AK083575.1 | - | 1 - 2 | 756 | 103 | |
| AK143943.1 | - | 2 - 3 | 547 | 103 | ||
| BC138097.1 | - | 1 - 2 | 689 | 8861116 | 103 | |
| BC138098.1 | - | 1 - 2 | 692 | 8861117 | 103 | |
| M22115.1 | - | 1 - 2 | 724 | 103 | ||
More Information
Set Membership: |
|

Hoxa1
Genomic Map