Assay Details
| Gene Symbol: Klk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010639.7 | NP_034769.4 | 3 - 4 | 535 | 82 | |
| GenBank mRNA | AK002278.1 | - | 3 - 4 | 536 | 82 | |
| BC010754.1 | - | 3 - 4 | 509 | 4240906 | 82 | |
| BC027736.1 | - | 3 - 4 | 513 | 4922971 | 82 | |
| BC037125.1 | - | 284 | 82 | |||
| BC053697.1 | - | 3 - 4 | 513 | 6529043 | 82 | |
| D10464.1 | - | 3 - 4 | 522 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klk1
Genomic Map