Assay Details
| Gene Symbol: Krt27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010666.1 | NP_034796.1 | 1 - 2 | 437 | 86 | |
| GenBank mRNA | AB013607.1 | - | 1 - 2 | 437 | 86 | |
| AK077384.1 | - | 1 - 2 | 449 | 86 | ||
| BC140996.1 | - | 1 - 2 | 461 | 9055608 | 86 | |
| BC140997.1 | - | 1 - 2 | 461 | 9055609 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Krt27
Genomic Map