Assay Details
| Gene Symbol: Lasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010688.4 | NP_034818.1 | 5 - 6 | 641 | 101 | |
| GenBank mRNA | AK034404.1 | - | 5 - 6 | 642 | 101 | |
| AK049199.1 | - | 5 - 6 | 642 | 101 | ||
| AK078445.1 | - | 5 - 6 | 642 | 101 | ||
| AK148858.1 | - | 5 - 6 | 644 | 101 | ||
| AK154656.1 | - | 5 - 6 | 642 | 101 | ||
| AK168521.1 | - | 5 - 6 | 642 | 101 | ||
| AK169381.1 | - | 5 - 6 | 642 | 101 | ||
| BC010840.1 | - | 5 - 6 | 626 | 3490532 | 101 | |
| CT010288.1 | - | 5 - 6 | 519 | 101 | ||
| U58882.1 | - | 3 - 4 | 345 | 101 | ||
| X96973.1 | - | 5 - 6 | 583 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lasp1
Genomic Map