Assay Details
| Gene Symbol: Ndufa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010886.2 | NP_035016.1 | 3 - 4 | 277 | 109 | |
| GenBank mRNA | AK005084.1 | - | 3 - 4 | 282 | 109 | |
| AK008357.1 | - | 3 - 4 | 274 | 109 | ||
| AK008506.1 | - | 3 - 4 | 268 | 109 | ||
| AK076036.1 | - | 3 - 4 | 278 | 109 | ||
| BC011114.1 | - | 3 - 4 | 240 | 3483028 | 109 | |
| U59509.1 | - | 3 - 4 | 175 | 109 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24239731 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24239732 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24239734 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24239733 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552832 | NM_010886.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552832 | AK005084.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552832 | AK008357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552832 | AK008506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552832 | AK076036.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552832 | NM_010886.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552832 | AK005084.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552832 | AK008357.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552832 | AK008506.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P552832 | AK076036.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554076 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116052 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 733987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69840 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30754 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173806 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552832 | NM_010886.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552832 | AK005084.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552832 | AK008357.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552832 | AK008506.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552832 | AK076036.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552832 | NM_010886.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552832 | AK005084.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552832 | AK008357.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552832 | AK008506.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P552832 | AK076036.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239770 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239765 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239724 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239719 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239771 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24239753 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173806 | - | - |
| Whole Mouse Genome G4122A | A_52_P552832 | NM_010886.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P552832 | AK005084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P552832 | AK008357.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P552832 | AK008506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P552832 | AK076036.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P552832 | NM_010886.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P552832 | AK005084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P552832 | AK008357.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P552832 | AK008506.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P552832 | AK076036.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101500 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173806 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05748850 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05390395 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05748856 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05748851 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05390385 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24239731 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24239732 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24239734 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24239733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24239720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05390395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05030007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05320858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239776 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239729 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05600207 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05748856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05145474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05605108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05390397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05390384 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05011636 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05748850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05320859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24239739 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10543138 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543137 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543136 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543139 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10543135 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424085_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10543134 | - | - |
| Mouse Genome 430 2.0 Array | 1424085_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424085_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17992_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17992_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600014657.mm.1 | TC0600001862.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600014657.mm.1 | NM_010886.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014657.mm.1 | NM_010886.2 | Refseq |
| Mouse Expression Set 430 | 1424085_PM_at | - | - |
| Mouse Expression Set 430 | 1424085_at | - | - |
| Murine Genome U74v2 Set | 160477_at | - | - |
| Murine Genome U74v2 Set | 160477_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424085_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424085_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424085_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424085_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6860364 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6860364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006860364 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2666062 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2666062 | - | - |
| mouse-6 v1.1 (Illumina) | scl017992.3_16-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6860364 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl017992.3_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860364 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2666061 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2666061 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl017992.3_16-S | - | - |

Ndufa4
Genomic Map