Assay Details
| Gene Symbol: Spryd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025697.3 | NP_079973.1 | 4 - 5 | 754 | 95 | |
| GenBank mRNA | AK018153.1 | - | 4 - 5 | 755 | 95 | |
| AK018290.1 | - | 4 - 5 | 604 | 95 | ||
| AK137685.1 | - | 603 | 95 | |||
| AK167306.1 | - | 4 - 5 | 596 | 95 | ||
| AK169059.1 | - | 4 - 5 | 603 | 95 | ||
| BC037015.1 | - | 4 - 5 | 588 | 5344409 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spryd7
Genomic Map