Assay Details
| Gene Symbol: Arl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025859.3 | NP_080135.2 | 4 - 5 | 478 | 98 | |
| GenBank mRNA | AK009226.1 | - | 498 | 98 | ||
| AK010732.1 | - | 4 - 5 | 479 | 98 | ||
| AK018138.1 | - | 480 | 98 | |||
| AK088330.1 | - | 527 | 98 | |||
| AK167748.1 | - | 475 | 98 | |||
| AK208964.1 | - | 3 - 4 | 257 | 98 | ||
| AK210028.1 | - | 3 - 4 | 288 | 98 | ||
| AK210540.1 | - | 3 - 4 | 284 | 98 | ||
| AK213037.1 | - | 3 - 4 | 240 | 98 | ||
| AK215653.1 | - | 3 - 4 | 266 | 98 | ||
| AK216046.1 | - | 3 - 4 | 257 | 98 | ||
| AK216240.1 | - | 3 - 4 | 260 | 98 | ||
| BC116894.1 | - | 492 | 40126213 | 98 | ||
| BC116896.1 | - | 492 | 40126215 | 98 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Arl1
Genomic Map