Assay Details
| Gene Symbol: Rpl37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026069.3 | NP_080345.1 | 1 - 2 | 71 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_026069.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl37
Genomic Map