Assay Details
| Gene Symbol: Ankrd33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026153.3 | NP_080429.1 | 1 - 2 | 723 | 102 | |
| GenBank mRNA | AK002572.1 | - | 1 - 2 | 518 | 102 | |
| AK012871.1 | - | 1 - 2 | 531 | 102 | ||
| AK170894.1 | - | 1 - 2 | 497 | 102 | ||
More Information
Set Membership: |
|

Ankrd33b
Genomic Map