Assay Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026883.3 | NP_081159.1 | 1 - 2 | 79 | 109 | |
| GenBank mRNA | AK005211.1 | - | 1 - 2 | 471 | 109 | |
| AK150218.1 | - | 1 - 2 | 478 | 109 | ||
| BC019498.1 | - | 1 - 2 | 437 | 4460879 | 109 | |
| BC057658.1 | - | 1 - 2 | 118 | 3153717 | 109 | |
| BC061033.1 | - | 1 - 2 | 79 | 6590939 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005211.1, AK150218.1, BC019498.1, BC057658.1, BC061033.1, NM_026883.3 transcripts.
Set Membership: |
|

Fam216a
Genomic Map