Assay Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026883.3 | NP_081159.1 | 4 - 5 | 391 | 97 | |
| GenBank mRNA | AK005211.1 | - | 4 - 5 | 783 | 97 | |
| AK150218.1 | - | 4 - 5 | 790 | 97 | ||
| BC019498.1 | - | 4 - 5 | 749 | 4460879 | 97 | |
| BC057658.1 | - | 4 - 5 | 430 | 3153717 | 97 | |
| BC061033.1 | - | 4 - 5 | 391 | 6590939 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam216a
Genomic Map