Assay Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026883.3 | NP_081159.1 | 6 - 7 | 644 | 110 | |
| GenBank mRNA | AK005211.1 | - | 6 - 7 | 1036 | 110 | |
| AK182306.1 | - | 2 - 3 | 172 | 110 | ||
| BC019498.1 | - | 6 - 7 | 1002 | 4460879 | 110 | |
| BC057658.1 | - | 6 - 7 | 683 | 3153717 | 110 | |
| BC061033.1 | - | 6 - 7 | 644 | 6590939 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050005 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050006 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050009 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050007 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050003 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050004 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252784 | BC061033.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252784 | NM_026883.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252784 | AK005211.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252784 | BC019498.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252784 | BC057658.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252784 | BC061033.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252784 | NM_026883.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252784 | AK005211.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252784 | BC019498.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252784 | BC057658.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297423 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910474 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178225 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252784 | BC061033.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252784 | NM_026883.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252784 | AK005211.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252784 | BC019498.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252784 | BC057658.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252784 | BC061033.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252784 | NM_026883.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252784 | AK005211.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252784 | BC019498.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252784 | BC057658.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24050007 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178225 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05724402 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01207317 | - | - |
| Whole Mouse Genome G4122A | A_51_P252784 | BC061033.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252784 | NM_026883.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252784 | AK005211.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252784 | BC019498.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252784 | BC057658.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252784 | BC061033.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252784 | NM_026883.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252784 | AK005211.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252784 | BC019498.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252784 | BC057658.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117899 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117854 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178225 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04734018 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04914235 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04628111 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04914233 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04557982 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050003 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050004 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050007 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050006 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050009 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24050005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24050007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04268462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04484957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04824336 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049998 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04672610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04334069 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04401582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24050002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04914233 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04599177 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10533476 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533480 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533477 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533478 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533481 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533479 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533475 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533482 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424192_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10533474 | - | - |
| Mouse Genome 430 2.0 Array | 1424192_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435565_at | - | - |
| Mouse Genome 430 2.0 Array | 1424192_at | - | - |
| Mouse Genome 430 2.0 Array | 1435565_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68948_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68948_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500028676.mm.1 | TC0500003208.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028676.mm.1 | NM_026883.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028676.mm.1 | NM_026883.3 | Refseq |
| Mouse Expression Set 430 | 1424192_at | - | - |
| Mouse Expression Set 430 | 1424192_PM_at | - | - |
| Murine Genome U74v2 Set | 160749_at | - | - |
| Murine Genome U74v2 Set | 160749_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424192_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424192_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424192_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424192_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5690364 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5690364 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005690364 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2775285 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2775285 | - | - |
| mouse-6 v1.1 (Illumina) | scl26077.1_53-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5690364 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26077.1_53-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690364 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2973024 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2775285 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2973024 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl26077.1_53-S | - | - |

Fam216a
Genomic Map