Assay Details
| Gene Symbol: Avpi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027106.4 | NP_081382.1 | 1 - 2 | 118 | 67 | |
| GenBank mRNA | AK009243.1 | - | 1 - 2 | 119 | 67 | |
| BC027646.1 | - | 1 - 2 | 152 | 1344413 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009243.1, BC027646.1, NM_027106.4 transcripts.
Set Membership: |
|

Avpi1
Genomic Map