Assay Details
| Gene Symbol: Mtfmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027134.3 | NP_081410.2 | 2 - 3 | 437 | 99 | |
| GenBank mRNA | AK009430.1 | - | 2 - 3 | 438 | 99 | |
| AK133492.1 | - | 2 - 3 | 432 | 99 | ||
| BC019509.1 | - | 2 - 3 | 427 | 4456981 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26587671 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26587670 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26587672 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2129856 | BC019509.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2129856 | NM_027134.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2129856 | AK009430.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2129856 | AK133492.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 201431 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 778441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848421 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376121 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909720 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 623528 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19642 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 127716 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62276 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 90863 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175024 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175001 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2129856 | BC019509.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2129856 | NM_027134.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2129856 | AK009430.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2129856 | AK133492.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P542204 | BC019509.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P542204 | NM_027134.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P542204 | AK009430.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P542204 | AK133492.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587682 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26587660 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175001 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06821946 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175024 | - | - |
| Whole Mouse Genome G4122A | A_52_P542204 | BC019509.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P542204 | NM_027134.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P542204 | AK009430.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P542204 | AK133492.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104860 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175001 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175024 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07525396 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07564234 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07525395 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07525398 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26587672 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26587671 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26587670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26587770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587710 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587743 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07760392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07564237 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07525394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08013723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07824339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08013719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07564241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07900300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07760393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07889249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07525398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07525397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08049817 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26587693 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10586400 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586398 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586403 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586399 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586401 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586404 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10586402 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457005_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424908_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10586397 | - | - |
| Mouse Genome 430 2.0 Array | 1424908_at | - | - |
| Mouse Genome 430 2.0 Array | 1457005_at | - | - |
| Mouse Genome 430 2.0 Array | 1424908_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457005_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69606_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69606_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900006431.mm.1 | NM_027134.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006431.mm.1 | NM_027134.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006431.mm.1 | TC0900000844.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006428.mm.1 | NM_027134.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006428.mm.1 | NM_027134.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006428.mm.1 | TC0900000844.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006430.mm.1 | NM_027134.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006430.mm.1 | NM_027134.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006430.mm.1 | TC0900000844.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900006429.mm.1 | NM_027134.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006429.mm.1 | NM_027134.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900006429.mm.1 | TC0900000844.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424908_at | - | - |
| Mouse Expression Set 430 | 1424908_PM_at | - | - |
| Mouse Expression Set 430 | 1457005_PM_at | - | - |
| Mouse Expression Set 430 | 1457005_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424908_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424908_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424908_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424908_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2850484 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2680433 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002850484 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002680433 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2734667 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2715153 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734667 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2715153 | - | - |
| mouse-6 v1.1 (Illumina) | scl069606.1_13-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0069606.2_82-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2850484 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2680433 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0069606.2_82-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl069606.1_13-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2680433 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850484 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2715153 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2734667 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2715152 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2734667 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2715153 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2715152 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl069606.1_13-S | - | - |

Mtfmt
Genomic Map