Assay Details
| Gene Symbol: Wars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027462.4 | NP_081738.2 | 1 - 2 | 98 | 85 | |
| GenBank mRNA | AK017602.1 | - | 1 - 2 | 109 | 85 | |
| AK033774.1 | - | 96 | 85 | |||
| AK036902.1 | - | 1 - 2 | 118 | 85 | ||
| AK042715.1 | - | 1 - 2 | 99 | 85 | ||
| BC089583.1 | - | 1 - 2 | 120 | 6762967 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wars2
Genomic Map