Assay Details
| Gene Symbol: Wars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027462.4 | NP_081738.2 | 3 - 4 | 435 | 72 | |
| GenBank mRNA | AK017602.1 | - | 3 - 4 | 446 | 72 | |
| AK033774.1 | - | 433 | 72 | |||
| AK036902.1 | - | 3 - 4 | 455 | 72 | ||
| AK042715.1 | - | 3 - 4 | 436 | 72 | ||
| AK042964.1 | - | 3 - 4 | 436 | 72 | ||
| BC089583.1 | - | 3 - 4 | 457 | 6762967 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wars2
Genomic Map