Assay Details
| Gene Symbol: Wars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027462.4 | NP_081738.2 | 4 - 5 | 521 | 101 | |
| GenBank mRNA | AK042715.1 | - | 4 - 5 | 522 | 101 | |
| AK042964.1 | - | 4 - 5 | 522 | 101 | ||
| BC089583.1 | - | 4 - 5 | 543 | 6762967 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wars2
Genomic Map