Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100034375.mm.1 |
NM_027983.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034375.mm.1 |
NM_027983.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034375.mm.1 |
TC1100003748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034373.mm.1 |
NM_027983.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034373.mm.1 |
NM_027983.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034373.mm.1 |
TC1100003748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034376.mm.1 |
NM_027983.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034376.mm.1 |
NM_027983.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034376.mm.1 |
TC1100003748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034379.mm.1 |
NM_027983.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034379.mm.1 |
NM_027983.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034379.mm.1 |
TC1100003748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034381.mm.1 |
NM_027983.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034381.mm.1 |
NM_027983.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034381.mm.1 |
TC1100003748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034377.mm.1 |
NM_027983.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034377.mm.1 |
NM_027983.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034377.mm.1 |
TC1100003748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034380.mm.1 |
NM_027983.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034380.mm.1 |
NM_027983.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034380.mm.1 |
TC1100003748.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100034374.mm.1 |
NM_027983.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034374.mm.1 |
NM_027983.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100034374.mm.1 |
TC1100003748.mm.1 |
MTA TC |