Assay Details
| Gene Symbol: Fam101a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028443.2 | NP_082719.1 | 2 - 3 | 380 | 77 | |
| GenBank mRNA | AK014112.1 | - | 2 - 3 | 410 | 77 | |
| BC053444.1 | - | 2 - 3 | 380 | 6336076 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24064187 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24064189 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368285 | AK014112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368285 | BC053444.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368285 | NM_028443.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368285 | AK014112.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368285 | BC053444.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368285 | NM_028443.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261706 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227878 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604353 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17933 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295942 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133776 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164221 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P166520 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100644 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368285 | AK014112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368285 | BC053444.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368285 | NM_028443.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368285 | AK014112.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368285 | BC053444.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368285 | NM_028443.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368285 | AK014112.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368285 | BC053444.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368285 | NM_028443.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368285 | AK014112.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368285 | BC053444.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368285 | NM_028443.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064113 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064099 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064105 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064100 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064097 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064102 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064103 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24064136 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164221 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100644 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P166520 | - | - |
| Whole Mouse Genome G4122A | A_51_P368285 | AK014112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368285 | BC053444.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368285 | NM_028443.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368285 | AK014112.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368285 | BC053444.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P368285 | NM_028443.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P132267 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P166520 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164221 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100644 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04602481 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04422685 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04602479 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04602478 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04766688 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24064187 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24064189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24064128 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04644986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04961029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04766690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04849512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04949849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04961018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04602479 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04451807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064095 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04766687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04849509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064102 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04445783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04451812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04462432 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064096 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064108 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064177 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04462435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064111 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24064158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04206716 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10525882 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525884 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10525883 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453192_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441881_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10525880 | - | - |
| Mouse Genome 430 2.0 Array | 1441881_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1441881_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1453192_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453192_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73121_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73121_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500013218.mm.1 | TC0500001455.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500013218.mm.1 | NM_028443.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013218.mm.1 | NM_028443.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013217.mm.1 | TC0500001455.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500013217.mm.1 | NM_028443.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013217.mm.1 | NM_028443.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013216.mm.1 | TC0500001455.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500013216.mm.1 | NM_028443.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013216.mm.1 | NM_028443.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013214.mm.1 | TC0500001455.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500013214.mm.1 | NM_028443.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500013214.mm.1 | NM_028443.2 | Refseq |
| Mouse Expression Set 430 | 1453192_at | - | - |
| Mouse Expression Set 430 | 1441881_x_at | - | - |
| Mouse Expression Set 430 | 1441881_PM_x_at | - | - |
| Mouse Expression Set 430 | 1453192_PM_at | - | - |
| Murine Genome U74v2 Set | 167555_i_at | - | - |
| Murine Genome U74v2 Set | 167555_PM_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441881_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453192_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453192_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441881_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 458300 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5570040 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005570040 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244695 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244695 | - | - |
| mouse-6 v1.1 (Illumina) | scl27213.3_538-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5570040 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl27213.3_538-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5570040 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1244695 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244695 | - | - |

Fam101a
Genomic Map