Assay Details
| Gene Symbol: Ptchd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028474.1 | NP_082750.1 | 1 - 2 | 361 | 79 | |
| GenBank mRNA | AK014270.1 | - | 1 - 2 | 320 | 79 | |
| BC145294.1 | - | 1 - 2 | 171 | 9053848 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014270.1, BC145294.1, NM_028474.1 transcripts.
Set Membership: |
|

Ptchd4
Genomic Map