Assay Details
| Gene Symbol: Ptchd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028474.1 | NP_082750.1 | 2 - 3 | 945 | 99 | |
| GenBank mRNA | BC145294.1 | - | 2 - 3 | 755 | 9053848 | 99 |
| BC151166.1 | - | 1 - 2 | 637 | 9088055 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptchd4
Genomic Map