| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300014636.mm.1 |
NR_045380.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014636.mm.1 |
NR_045380.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014636.mm.1 |
TC0300001906.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300014635.mm.1 |
NR_045380.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014635.mm.1 |
NR_045380.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014635.mm.1 |
TC0300001906.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300014639.mm.1 |
NR_045380.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014639.mm.1 |
NR_045380.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014639.mm.1 |
TC0300001906.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300014637.mm.1 |
NR_045380.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014637.mm.1 |
NR_045380.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300014637.mm.1 |
TC0300001906.mm.1 |
MTA TC |