Assay Details
Gene Symbol: Lrrc48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029044.2 | NP_083320.1 | 9 - 10 | 1184 | 76 | |
GenBank mRNA | AK015430.1 | - | 9 - 10 | 1186 | 76 | |
BC018412.1 | - | 9 - 10 | 1197 | 3673669 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
