Assay Details
Gene Symbol: Rasd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029182.1 | NP_083458.1 | 1 - 2 | 136 | 104 | |
GenBank mRNA | BC036988.1 | - | 1 - 2 | 147 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_029182.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
