Assay Details
| Gene Symbol: Vmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029478.3 | NP_083754.2 | 1 - 2 | 85 | 92 | |
| GenBank mRNA | AB047550.1 | - | 1 - 2 | 97 | 92 | |
| AK016312.1 | - | 1 - 2 | 102 | 92 | ||
| AK043091.1 | - | 1 - 2 | 86 | 92 | ||
| AK049559.1 | - | 1 - 2 | 78 | 92 | ||
| AK077443.1 | - | 1 - 2 | 85 | 92 | ||
| AK159468.1 | - | 1 - 2 | 100 | 92 | ||
| AK167173.1 | - | 1 - 2 | 94 | 92 | ||
| AK167202.1 | - | 1 - 2 | 94 | 92 | ||
| AX775963.1 | - | 1 - 2 | 55 | 92 | ||
| BC004013.1 | - | 1 - 2 | 79 | 3493738 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB047550.1,AK043091.1,AK077443.1,AK159468.1,AX775963.1,BC004013.1,NM_029478.3 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vmp1
Genomic Map