Assay Details
| Gene Symbol: Vmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029478.3 | NP_083754.2 | 2 - 3 | 185 | 107 | |
| GenBank mRNA | AB047550.1 | - | 2 - 3 | 197 | 107 | |
| AB076609.1 | - | 1 - 2 | 76 | 107 | ||
| AK016312.1 | - | 2 - 3 | 202 | 107 | ||
| AK043091.1 | - | 2 - 3 | 186 | 107 | ||
| AK049559.1 | - | 2 - 3 | 178 | 107 | ||
| AK050796.1 | - | 1 - 2 | 54 | 107 | ||
| AK077443.1 | - | 2 - 3 | 185 | 107 | ||
| AK159468.1 | - | 2 - 3 | 200 | 107 | ||
| AK167173.1 | - | 2 - 3 | 194 | 107 | ||
| AK167202.1 | - | 2 - 3 | 194 | 107 | ||
| AX775963.1 | - | 2 - 3 | 155 | 107 | ||
| BC004013.1 | - | 2 - 3 | 179 | 3493738 | 107 | |
More Information
Set Membership: |
|

Vmp1
Genomic Map