Assay Details
| Gene Symbol: Vmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029478.3 | NP_083754.2 | 5 - 6 | 516 | 106 | |
| GenBank mRNA | AB047550.1 | - | 5 - 6 | 528 | 106 | |
| AB076609.1 | - | 4 - 5 | 407 | 106 | ||
| AK016312.1 | - | 5 - 6 | 533 | 106 | ||
| AK043091.1 | - | 5 - 6 | 517 | 106 | ||
| AK049559.1 | - | 5 - 6 | 509 | 106 | ||
| AK050796.1 | - | 4 - 5 | 385 | 106 | ||
| AK077443.1 | - | 5 - 6 | 516 | 106 | ||
| AK159468.1 | - | 5 - 6 | 531 | 106 | ||
| AK167173.1 | - | 5 - 6 | 524 | 106 | ||
| AK167202.1 | - | 5 - 6 | 524 | 106 | ||
| AX775963.1 | - | 5 - 6 | 486 | 106 | ||
| BC004013.1 | - | 5 - 6 | 510 | 3493738 | 106 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vmp1
Genomic Map