Assay Details
| Gene Symbol: Vmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029478.3 | NP_083754.2 | 8 - 9 | 905 | 108 | |
| GenBank mRNA | AB047550.1 | - | 8 - 9 | 917 | 108 | |
| AB076609.1 | - | 7 - 8 | 796 | 108 | ||
| AK016312.1 | - | 8 - 9 | 922 | 108 | ||
| AK043091.1 | - | 8 - 9 | 906 | 108 | ||
| AK049559.1 | - | 8 - 9 | 898 | 108 | ||
| AK077443.1 | - | 8 - 9 | 905 | 108 | ||
| AK159468.1 | - | 8 - 9 | 920 | 108 | ||
| AK167173.1 | - | 8 - 9 | 913 | 108 | ||
| AK167202.1 | - | 8 - 9 | 913 | 108 | ||
| AX775963.1 | - | 8 - 9 | 875 | 108 | ||
| BC004013.1 | - | 8 - 9 | 899 | 3493738 | 108 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vmp1
Genomic Map