Assay Details
| Gene Symbol: Prl2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031191.1 | NP_112468.1 | 1 - 2 | 36 | 82 | |
| GenBank mRNA | BC056203.1 | - | 1 - 2 | 86 | 30014401 | 82 |
| BC064771.1 | - | 1 - 2 | 92 | 30049431 | 82 | |
| BC080826.1 | - | 1 - 2 | 101 | 30438424 | 82 | |
| BC100300.1 | - | 1 - 2 | 71 | 30715342 | 82 | |
| BC145439.1 | - | 1 - 2 | 64 | 9053997 | 82 | |
| K02245.1 | - | 1 - 2 | 36 | 82 | ||
| X75557.1 | - | 1 - 2 | 40 | 82 | ||
| Gene Symbol: Prl2c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC132514.1 | - | 1 - 2 | 64 | 40130791 | 82 |
| BC132516.1 | - | 1 - 2 | 64 | 40130793 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC056203.1, BC064771.1, BC080826.1, BC100300.1, BC132514.1, BC132516.1, BC145439.1, K02245.1, NM_031191.1, X75557.1 transcripts.
Set Membership: |
|

Prl2c2
Genomic Map