Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600017527.mm.1 |
TC0600002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017527.mm.1 |
NM_031247.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017527.mm.1 |
NM_031247.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017529.mm.1 |
TC0600002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017529.mm.1 |
NM_031247.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017529.mm.1 |
NM_031247.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017530.mm.1 |
TC0600002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017530.mm.1 |
NM_031247.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017530.mm.1 |
NM_031247.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017524.mm.1 |
TC0600002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017524.mm.1 |
NM_031247.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017524.mm.1 |
NM_031247.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017525.mm.1 |
TC0600002229.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017525.mm.1 |
NM_031247.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017525.mm.1 |
NM_031247.3 |
Refseq |