Assay Details
| Gene Symbol: Trem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031254.3 | NP_112544.1 | 3 - 4 | 637 | 98 | |
| GenBank mRNA | AF213458.1 | - | 3 - 4 | 548 | 98 | |
| AK039477.1 | - | 3 - 4 | 542 | 98 | ||
| AK171358.1 | - | 2460 | 98 | |||
| AY024348.1 | - | 3 - 4 | 557 | 98 | ||
| AY024349.1 | - | 3 - 4 | 482 | 98 | ||
| BC032959.1 | - | 3 - 4 | 526 | 1348066 | 98 | |
| BC033485.1 | - | 3 - 4 | 548 | 4953284 | 98 | |
| BC052784.1 | - | 3 - 4 | 646 | 5057491 | 98 | |
More Information
Set Membership: |
|

Trem2
Genomic Map