Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800002772.mm.1 |
NM_033579.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002772.mm.1 |
NM_033579.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002772.mm.1 |
TC1800001741.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033575.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033575.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033576.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033576.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033577.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033577.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033578.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033578.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033579.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033579.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033580.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033580.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033581.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033581.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033582.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033582.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033583.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033583.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033584.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033584.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033586.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033586.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033587.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033587.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033588.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033588.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033589.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033589.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033590.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033590.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033591.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033591.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033592.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033592.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033593.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033593.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033594.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033594.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033595.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
NM_033595.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800002789.mm.1 |
TC1800001741.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800002771.mm.1 |
TC1800001741.mm.1 |
MTA TC |