Assay Details
| Gene Symbol: Slc15a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053079.2 | NP_444309.2 | 15 - 16 | 1178 | 97 | |
| GenBank mRNA | AF205540.1 | - | 15 - 16 | 1177 | 97 | |
| AK085471.1 | - | 13 - 14 | 2285 | 97 | ||
| AK136428.1 | - | 15 - 16 | 1179 | 97 | ||
| BC116248.1 | - | 15 - 16 | 1168 | 40093541 | 97 | |
| BC116249.1 | - | 15 - 16 | 1168 | 40093542 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc15a1
Genomic Map