Assay Details
| Gene Symbol: Slc15a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053079.2 | NP_444309.2 | 18 - 19 | 1502 | 86 | |
| GenBank mRNA | AK136428.1 | - | 18 - 19 | 1503 | 86 | |
| BC116248.1 | - | 18 - 19 | 1492 | 40093541 | 86 | |
| BC116249.1 | - | 18 - 19 | 1492 | 40093542 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc15a1
Genomic Map