Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400020371.mm.1 |
NM_053079.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020371.mm.1 |
NM_053079.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020371.mm.1 |
TC1400002738.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400020373.mm.1 |
NM_053079.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020373.mm.1 |
NM_053079.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020373.mm.1 |
TC1400002738.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400020372.mm.1 |
NM_053079.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020372.mm.1 |
NM_053079.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020372.mm.1 |
TC1400002738.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400020377.mm.1 |
NM_053079.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020377.mm.1 |
NM_053079.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400020377.mm.1 |
TC1400002738.mm.1 |
MTA TC |