Assay Details
| Gene Symbol: Btnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079835.2 | NP_524574.1 | 5 - 6 | 986 | 97 | |
| GenBank mRNA | AK136521.1 | - | 5 - 6 | 988 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Btnl2
Genomic Map